Product Details
- SNP ID
-
rs191618578
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:26046823 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTGTTGGGGGCGCCCGGGCCCCT[A/G]AGTTGGCGCTGGAGCTGGAACTGGC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SHISA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74965018] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SHISA2
- Gene Name
- shisa family member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001007538.1 |
663 |
Missense Mutation |
TCA,TTA |
S193L |
NP_001007539.1 |
View Full Product Details