Product Details

SNP ID
rs191515346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126438412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGGTTGATAGTGCTTCCTGGG[A/T]ACTGGGTGGCCTCCGGTCCAGCAGG
Phenotype
MIM: 615746
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CFAP100 PubMed Links

Gene Details

Gene
CFAP100
Gene Name
cilia and flagella associated protein 100
There are no transcripts associated with this gene.

Gene
LOC105374089
Gene Name
uncharacterized LOC105374089
There are no transcripts associated with this gene.

Gene
ZXDC
Gene Name
ZXD family zinc finger C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040653.3 2339 Intron NP_001035743.1
NM_025112.4 2339 Missense Mutation TAC,TTC Y847F NP_079388.3
XM_005247757.3 2339 Intron XP_005247814.1
XM_006713741.2 2339 Missense Mutation TAC,TTC Y758F XP_006713804.1
XM_011513119.2 2339 Missense Mutation TAC,TTC Y790F XP_011511421.1

View Full Product Details