Product Details

SNP ID
rs191533610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:20364327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTTGGGCGCCAGCGGAGAAAC[C/T]GGCAGAGTGCTCTTAAAGGAAATCC
Phenotype
MIM: 605628
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTATIP2 PubMed Links

Gene Details

Gene
HTATIP2
Gene Name
HIV-1 Tat interactive protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098520.1 156 Silent Mutation ACC,ACT T64T NP_001091990.1
NM_001098521.1 156 Silent Mutation ACC,ACT T30T NP_001091991.1
NM_001098522.1 156 Silent Mutation ACC,ACT T30T NP_001091992.1
NM_001098523.1 156 Silent Mutation ACC,ACT T30T NP_001091993.1
NM_006410.4 156 Silent Mutation ACC,ACT T30T NP_006401.3

View Full Product Details