Product Details

SNP ID
rs191751735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35739786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGCGCCTGGGGAGGACAGATG[C/T]GAGGAGGAAGGGGTGTGGGTGTTGG
Phenotype
MIM: 614143 MIM: 606834 MIM: 607632 MIM: 601080
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGFLR1 PubMed Links

Gene Details

Gene
IGFLR1
Gene Name
IGF like family receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024660.2 743 Silent Mutation NP_078936.1
Gene
KMT2B
Gene Name
lysine methyltransferase 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014727.2 743 Intron NP_055542.1
XM_011527561.2 743 Intron XP_011525863.2
XM_011527562.2 743 Intron XP_011525864.1
XM_017027544.1 743 Intron XP_016883033.1
XM_017027545.1 743 Intron XP_016883034.1
XM_017027546.1 743 Intron XP_016883035.1
Gene
LIN37
Gene Name
lin-37 DREAM MuvB core complex component
There are no transcripts associated with this gene.

Gene
PSENEN
Gene Name
presenilin enhancer gamma-secretase subunit
There are no transcripts associated with this gene.

Gene
U2AF1L4
Gene Name
U2 small nuclear RNA auxiliary factor 1 like 4
There are no transcripts associated with this gene.

View Full Product Details