Product Details

SNP ID
rs192022515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:18270030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGACCCTGGCATACACTCCGGGG[C/T]GATTAGGCAGGGCACACTTGTATCC
Phenotype
MIM: 607247 MIM: 606635
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHODL PubMed Links

Gene Details

Gene
CHODL
Gene Name
chondrolectin
There are no transcripts associated with this gene.

Gene
TMPRSS15
Gene Name
transmembrane protease, serine 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002772.2 3194 Missense Mutation CAC,CGC H1000R NP_002763.2
XM_011529654.1 3194 Missense Mutation CAC,CGC H1045R XP_011527956.1
XM_011529655.1 3194 Missense Mutation CAC,CGC H1045R XP_011527957.1
XM_011529656.2 3194 Missense Mutation CAC,CGC H1045R XP_011527958.1
XM_011529657.1 3194 Missense Mutation CAC,CGC H1030R XP_011527959.1
XM_011529658.1 3194 Missense Mutation CAC,CGC H1018R XP_011527960.1
XM_011529659.1 3194 Missense Mutation CAC,CGC H1015R XP_011527961.1

View Full Product Details