Product Details

SNP ID
rs191630054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37800932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCGGAGCCGGGGCTGAGGAGCC[A/G]ACGCCCCACGAGGGGCAGGCAGCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF10 PubMed Links

Gene Details

Gene
DCAF10
Gene Name
DDB1 and CUL4 associated factor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286810.1 168 Intron NP_001273739.1
NM_024345.4 168 Silent Mutation CCA,CCG P22P NP_077321.3
XM_005251577.3 168 Silent Mutation CCA,CCG P22P XP_005251634.1
XM_005251578.3 168 Silent Mutation CCA,CCG P22P XP_005251635.1
XM_017015128.1 168 Silent Mutation CCA,CCG P22P XP_016870617.1
XM_017015129.1 168 Intron XP_016870618.1
Gene
LOC105376037
Gene Name
uncharacterized LOC105376037
There are no transcripts associated with this gene.

View Full Product Details