Product Details

SNP ID
rs191663634
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:156293154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGGCCACTGCCATGATGCCCA[C/G]GACTAGTGGGGACAAGCCGTCCACT
Phenotype
MIM: 610962 MIM: 615531
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLMP PubMed Links

Gene Details

Gene
GLMP
Gene Name
glycosylated lysosomal membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256604.1 814 Silent Mutation CTG,GTG L322V NP_001243533.1
NM_001256605.1 814 Silent Mutation CTG,GTG L285V NP_001243534.1
NM_001256608.1 814 Missense Mutation CCT,CGT P285R NP_001243537.1
NM_001256609.1 814 Silent Mutation CTG,GTG L290V NP_001243538.1
NM_144580.2 814 Silent Mutation CTG,GTG L371V NP_653181.1
XM_011509117.2 814 Silent Mutation CTG,GTG L194V XP_011507419.1
XM_017000149.1 814 UTR 3 XP_016855638.1
Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

Gene
TMEM79
Gene Name
transmembrane protein 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032323.2 814 Intron NP_115699.1
Gene
VHLL
Gene Name
von Hippel-Lindau tumor suppressor like
There are no transcripts associated with this gene.

View Full Product Details