Product Details

SNP ID
rs192535892
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185164045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCTTACTTAATGGGCTATATTG[C/G]CCAAGAGTGGATCTGGCCCGTCTTG
Phenotype
MIM: 616047
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CFAP97 PubMed Links

Gene Details

Gene
CFAP97
Gene Name
cilia and flagella associated protein 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292033.1 1602 Intron NP_001278962.1
NM_020827.2 1602 Silent Mutation GGC,GGG G485G NP_065878.1
XM_017008483.1 1602 Silent Mutation GGC,GGG G485G XP_016863972.1
XM_017008484.1 1602 Silent Mutation GGC,GGG G485G XP_016863973.1
XM_017008485.1 1602 Silent Mutation GGC,GGG G485G XP_016863974.1
XM_017008486.1 1602 Silent Mutation GGC,GGG G485G XP_016863975.1

View Full Product Details