Product Details

SNP ID
rs192362382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32692281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTTTCCCGTCGCTGCCCGACAC[C/G]GATGACCACAAGACCGACAACACTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NUDT19 PubMed Links

Gene Details

Gene
NUDT19
Gene Name
nudix hydrolase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105570.1 675 Silent Mutation ACC,ACG T107T NP_001099040.1
XM_017026806.1 675 Silent Mutation ACC,ACG T107T XP_016882295.1

View Full Product Details