Product Details

SNP ID
rs192361570
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47656988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTCCTCAGCTGGTGGCCAATGT[C/T]ACAAACCCCAACAGCACAGAGCACA
Phenotype
MIM: 602738
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KPNB1 PubMed Links

Gene Details

Gene
KPNB1
Gene Name
karyopherin subunit beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276453.1 493 UTR 5 NP_001263382.1
NM_002265.5 493 Silent Mutation GTC,GTT V137V NP_002256.2

View Full Product Details