Product Details

SNP ID
rs192381116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58369283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGCCCCGGAGGCACCCTGGGCA[A/T]CGGCCTTGGGGAGTCCTCCCTGCCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105372483 PubMed Links

Gene Details

Gene
LOC105372483
Gene Name
uncharacterized LOC105372483
There are no transcripts associated with this gene.

Gene
ZNF497
Gene Name
zinc finger protein 497
There are no transcripts associated with this gene.

Gene
ZNF837
Gene Name
zinc finger protein 837
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138466.1 1039 Missense Mutation GAT,GTT D17V NP_612475.1
XM_017026242.1 1039 Missense Mutation GAT,GTT D17V XP_016881731.1

View Full Product Details