Product Details

SNP ID
rs192395872
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:160880613 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCCATAGGGTGAGTAATAAGA[A/T]GCTGTTTTCTGGGCGTCGCCAAAAT
Phenotype
MIM: 609873
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ITLN1 PubMed Links

Gene Details

Gene
ITLN1
Gene Name
intelectin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017625.2 776 Silent Mutation GCA,GCT A220A NP_060095.2

View Full Product Details