Product Details

SNP ID
rs192509496
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:248521939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACCATGAGCTACGGTGGCTGTG[A/T]GGCCCAGCTCTATGTGGCCATGGGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2G6 PubMed Links

Gene Details

Gene
OR2G6
Gene Name
olfactory receptor family 2 subfamily G member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013355.1 293 Missense Mutation GAG,GTG E98V NP_001013373.1
Gene
OR2T10
Gene Name
olfactory receptor family 2 subfamily T member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004693.1 293 Intron NP_001004693.1
XM_017000273.1 293 Intron XP_016855762.1

View Full Product Details