Product Details

SNP ID
rs192794351
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:248239240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTGGAACTCAGATATTCTTCTA[A/T]GTGTCCCTGCTTGGAGCTGAATGTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2M4 PubMed Links

Gene Details

Gene
OR2M4
Gene Name
olfactory receptor family 2 subfamily M member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017504.1 312 Silent Mutation TAA,TAT *104Y NP_059974.1

View Full Product Details