Product Details

SNP ID
rs192837663
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:103526250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTCTGTGCGGGTCCTGTCAGA[A/T]AAGGGGCGACTGGTAAGGGCGCGCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928436 PubMed Links

Gene Details

Gene
LOC101928436
Gene Name
uncharacterized LOC101928436
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286997.1 Intron NP_001273926.1
Gene
RNPC3
Gene Name
RNA binding region (RNP1, RRM) containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017619.3 Intron NP_060089.1

View Full Product Details