Product Details

SNP ID
rs192876608
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65947001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGAGCAAGAAAGGACAGGGCT[A/C]TGAAGAGTCTGAAGAGTAAGCAGCA
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
TSGA10IP PubMed Links

Gene Details

Gene
TSGA10IP
Gene Name
testis specific 10 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152762.2 500 Missense Mutation TAT,TCT Y90S NP_689975.2
XM_011544887.2 500 Missense Mutation TAT,TCT Y90S XP_011543189.1
XM_011544888.2 500 UTR 5 XP_011543190.1

View Full Product Details