Product Details

SNP ID
rs192942360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:16273264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCACCCCTGGCTCTACCCCGTCC[C/T]GTGGCTGCTGTAGTCAAAGACTCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF16B PubMed Links

Gene Details

Gene
KIF16B
Gene Name
kinesin family member 16B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199865.1 3974 Missense Mutation AGG,GGG R1264G NP_001186794.1
NM_001199866.1 3974 Intron NP_001186795.1
NM_024704.4 3974 Missense Mutation AGG,GGG R1315G NP_078980.3
XM_005260750.3 3974 Intron XP_005260807.1
XM_005260751.4 3974 Intron XP_005260808.1
XM_005260752.3 3974 Intron XP_005260809.1
XM_005260753.3 3974 Missense Mutation AGG,GGG R1326G XP_005260810.1
XM_005260754.3 3974 Missense Mutation AGG,GGG R1296G XP_005260811.1
XM_005260755.3 3974 Missense Mutation AGG,GGG R1275G XP_005260812.1
XM_006723588.3 3974 Missense Mutation AGG,GGG R1285G XP_006723651.1
XM_017027926.1 3974 Missense Mutation AGG,GGG R1274G XP_016883415.1

View Full Product Details