Product Details

SNP ID
rs384845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41054520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTCTATGGTAGGAGTATTGAG[C/T]TTATTAGAAAGCAGACAACATGATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP2-1 PubMed Links

Gene Details

Gene
KRTAP2-1
Gene Name
keratin associated protein 2-1
There are no transcripts associated with this gene.

Gene
KRTAP2-2
Gene Name
keratin associated protein 2-2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033032.2 692 UTR 3 NP_149021.2
Gene
KRTAP2-3
Gene Name
keratin associated protein 2-3
There are no transcripts associated with this gene.

View Full Product Details