Product Details

SNP ID
rs437102
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:12242015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCAAGGGGTCAGCAAAGAGACC[A/C]GCCGGCCACATCTGGCCTGCAGACT
Phenotype
MIM: 608877
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
VPS13D PubMed Links

Gene Details

Gene
VPS13D
Gene Name
vacuolar protein sorting 13 homolog D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015378.3 Intron NP_056193.2
NM_018156.3 Intron NP_060626.2

View Full Product Details