Product Details

SNP ID
rs6000939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37916758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGTGGCTGTGGGAAATCAGAAA[A/G]GGGTTGGGGAGTAATGTCAGGATGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MICALL1 PubMed Links

Gene Details

Gene
MICALL1
Gene Name
MICAL like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033386.3 Intron NP_203744.1
XM_005261791.3 Intron XP_005261848.1
XM_005261792.3 Intron XP_005261849.1
XM_011530471.2 Intron XP_011528773.1
XM_011530472.2 Intron XP_011528774.1
XM_011530473.2 Intron XP_011528775.1
XM_011530474.2 Intron XP_011528776.1
XM_011530475.2 Intron XP_011528777.1
XM_011530476.2 Intron XP_011528778.1
XM_017029026.1 Intron XP_016884515.1

View Full Product Details