Product Details

SNP ID
rs5970393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152902182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGCGGCAGAGCTCTGAAGCCAC[A/G]CCCTGGGTCAGAGAGGAGACATAGC
Phenotype
MIM: 300381
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF185 PubMed Links

Gene Details

Gene
ZNF185
Gene Name
zinc finger protein 185 (LIM domain)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178106.1 Intron NP_001171577.1
NM_001178107.1 Intron NP_001171578.1
NM_001178108.1 Intron NP_001171579.1
NM_001178109.1 Intron NP_001171580.1
NM_001178110.1 Intron NP_001171581.1
NM_001178113.1 Intron NP_001171584.1
NM_001178115.1 Intron NP_001171586.1
NM_007150.3 Intron NP_009081.2
XM_005274730.2 Intron XP_005274787.1
XM_005274731.2 Intron XP_005274788.1
XM_005274735.2 Intron XP_005274792.1
XM_005274737.2 Intron XP_005274794.1
XM_005274738.2 Intron XP_005274795.1
XM_005274740.2 Intron XP_005274797.1
XM_005274741.2 Intron XP_005274798.1
XM_005274742.2 Intron XP_005274799.1
XM_005274744.2 Intron XP_005274801.1
XM_005274745.2 Intron XP_005274802.1
XM_005274746.3 Intron XP_005274803.1
XM_011531194.1 Intron XP_011529496.1
XM_011531195.1 Intron XP_011529497.1
XM_017029821.1 Intron XP_016885310.1
XM_017029822.1 Intron XP_016885311.1
XM_017029823.1 Intron XP_016885312.1
XM_017029824.1 Intron XP_016885313.1
XM_017029825.1 Intron XP_016885314.1
XM_017029826.1 Intron XP_016885315.1
XM_017029827.1 Intron XP_016885316.1
XM_017029828.1 Intron XP_016885317.1
XM_017029829.1 Intron XP_016885318.1
XM_017029830.1 Intron XP_016885319.1
XM_017029831.1 Intron XP_016885320.1
XM_017029832.1 Intron XP_016885321.1
XM_017029833.1 Intron XP_016885322.1
XM_017029834.1 Intron XP_016885323.1
XM_017029835.1 Intron XP_016885324.1

View Full Product Details