Product Details

SNP ID
rs6094095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45109654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCATTCTGGATGGTTGGGCTGC[C/T]TGAGGGAGAGGTCAGGGTCTGCCTG
Phenotype
MIM: 602905 MIM: 605161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNS1 PubMed Links

Gene Details

Gene
KCNS1
Gene Name
potassium voltage-gated channel modifier subfamily S member 1
There are no transcripts associated with this gene.

Gene
WFDC5
Gene Name
WAP four-disulfide core domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145652.3 816 UTR 3 NP_663627.1
XM_011528601.1 816 UTR 3 XP_011526903.1
XM_011528602.1 816 UTR 3 XP_011526904.1

View Full Product Details