Product Details

SNP ID
rs6724297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:207766273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTATTCAAGACACAACGATGGTGC[A/T]GTTAGGAGTGAGAATTATTACAAGC
Phenotype
MIM: 601723
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FZD5 PubMed Links

Gene Details

Gene
FZD5
Gene Name
frizzled class receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003468.3 2877 UTR 3 NP_003459.2

View Full Product Details