Product Details

SNP ID
rs6649210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:124381900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATTTTCAACATTTTGCTAAATG[C/T]TTTTTAGAAGGACTTTTAATTACAG
Phenotype
MIM: 300490 MIM: 300588
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH2D1A PubMed Links

Gene Details

Gene
SH2D1A
Gene Name
SH2 domain containing 1A
There are no transcripts associated with this gene.

Gene
TENM1
Gene Name
teneurin transmembrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163278.1 Intron NP_001156750.1
NM_001163279.1 Intron NP_001156751.1
NM_014253.3 Intron NP_055068.2
XM_011531230.2 Intron XP_011529532.1
XM_011531236.2 Intron XP_011529538.2
XM_011531237.2 Intron XP_011529539.1
XM_017029208.1 Intron XP_016884697.1
XM_017029209.1 Intron XP_016884698.1
XM_017029210.1 Intron XP_016884699.1
XM_017029211.1 Intron XP_016884700.1
XM_017029212.1 Intron XP_016884701.1
XM_017029213.1 Intron XP_016884702.1
XM_017029214.1 Intron XP_016884703.1
XM_017029215.1 Intron XP_016884704.1
XM_017029216.1 Intron XP_016884705.1

View Full Product Details