Product Details

SNP ID
rs7326661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:24932043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGGCGCCTTCTGGCGACGGTCC[C/G]TGCTAGGGACGCTGCGCTCTCCGAG
Phenotype
MIM: 609279
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CENPJ PubMed Links

Gene Details

Gene
CENPJ
Gene Name
centromere protein J
There are no transcripts associated with this gene.

Gene
TPTE2P1
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details