Product Details

SNP ID
rs748097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2516764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCCTGCAGAGGGGAAGACTAGG[A/G]TCCAAGAAGCCCATGAAGCCATGCG
Phenotype
MIM: 602576
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LFNG PubMed Links

Gene Details

Gene
LFNG
Gene Name
LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040167.1 Intron NP_001035257.1
NM_001040168.1 Intron NP_001035258.1
NM_001166355.1 Intron NP_001159827.1
NM_002304.2 Intron NP_002295.1

View Full Product Details