Product Details

SNP ID
rs7592617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68720775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAACACAACTTCCTTTCACCTGG[G/T]GAGTCTAAGCTGATCAAACTGCATG
Phenotype
MIM: 610587
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP25 PubMed Links

Gene Details

Gene
ARHGAP25
Gene Name
Rho GTPase activating protein 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007231.2 Intron NP_001007232.2
NM_001166276.1 Intron NP_001159748.1
NM_001166277.1 Intron NP_001159749.1
NM_014882.2 Intron NP_055697.1
XM_005264675.2 Intron XP_005264732.1
XM_005264676.2 Intron XP_005264733.1
XM_011533206.2 Intron XP_011531508.1
XM_011533207.2 Intron XP_011531509.1
XM_011533208.2 Intron XP_011531510.1
XM_011533209.2 Intron XP_011531511.1
XM_011533210.2 Intron XP_011531512.1
XM_017005423.1 Intron XP_016860912.1
XM_017005424.1 Intron XP_016860913.1
XM_017005425.1 Intron XP_016860914.1
XM_017005426.1 Intron XP_016860915.1
XM_017005427.1 Intron XP_016860916.1
XM_017005428.1 Intron XP_016860917.1

View Full Product Details