Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001297640.1 | 3236 | UTR 3 | NP_001284569.1 | ||
NM_001297642.1 | 3236 | UTR 3 | NP_001284571.1 | ||
NM_001297643.1 | 3236 | Intron | NP_001284572.1 | ||
NM_016546.3 | 3236 | UTR 3 | NP_057630.2 | ||
XM_017019397.1 | 3236 | UTR 3 | XP_016874886.1 | ||
XM_017019398.1 | 3236 | UTR 3 | XP_016874887.1 | ||
XM_017019399.1 | 3236 | Intron | XP_016874888.1 | ||
XM_017019400.1 | 3236 | Intron | XP_016874889.1 | ||
XM_017019401.1 | 3236 | Intron | XP_016874890.1 | ||
XM_017019402.1 | 3236 | Intron | XP_016874891.1 | ||
XM_017019403.1 | 3236 | Intron | XP_016874892.1 | ||
XM_017019404.1 | 3236 | Intron | XP_016874893.1 |