Product Details

SNP ID
rs7940811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:40107811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGAGGGCAAGTTGTACTTTTG[A/T]TACTTCAGTTGCACTCCAAGGAGTG
Phenotype
MIM: 608817
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LRRC4C PubMed Links

Gene Details

Gene
LRRC4C
Gene Name
leucine rich repeat containing 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258419.1 9706 Intron NP_001245348.1
NM_020929.2 9706 Intron NP_065980.1
XM_011520238.2 9706 UTR 3 XP_011518540.1
XM_011520239.2 9706 UTR 3 XP_011518541.1
XM_011520240.2 9706 UTR 3 XP_011518542.1
XM_011520241.2 9706 UTR 3 XP_011518543.1
XM_011520242.2 9706 UTR 3 XP_011518544.1
XM_011520243.2 9706 UTR 3 XP_011518545.1
XM_011520244.2 9706 UTR 3 XP_011518546.1
XM_017018070.1 9706 UTR 3 XP_016873559.1
XM_017018071.1 9706 UTR 3 XP_016873560.1
XM_017018072.1 9706 UTR 3 XP_016873561.1
XM_017018073.1 9706 UTR 3 XP_016873562.1
XM_017018074.1 9706 UTR 3 XP_016873563.1
XM_017018075.1 9706 UTR 3 XP_016873564.1
XM_017018076.1 9706 UTR 3 XP_016873565.1
XM_017018077.1 9706 UTR 3 XP_016873566.1
XM_017018078.1 9706 UTR 3 XP_016873567.1
XM_017018079.1 9706 UTR 3 XP_016873568.1

View Full Product Details