Product Details

SNP ID
rs7945066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:11352040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCAGAGGTCCAAGGGGTGAAGG[A/G]GTTGGCACTGAAGAAATCCCTGACA
Phenotype
MIM: 615136
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSNK2A3 PubMed Links

Gene Details

Gene
CSNK2A3
Gene Name
casein kinase 2 alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256686.1 Intron NP_001243615.1
Gene
GALNT18
Gene Name
polypeptide N-acetylgalactosaminyltransferase 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198516.2 Intron NP_940918.2
XM_006718224.2 Intron XP_006718287.1
XM_006718225.2 Intron XP_006718288.1
XM_011520068.2 Intron XP_011518370.1
XM_011520069.2 Intron XP_011518371.1
XM_011520070.2 Intron XP_011518372.1
XM_011520071.2 Intron XP_011518373.1

View Full Product Details