Product Details
- SNP ID
-
rs7987863
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:45120004 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTACTTGTTCGTTTCGTTACAGAC[C/T]TCCGGGTTTTGGAAACTGAGGTGCT
- Phenotype
-
MIM: 189969
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GTF2F2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9567531] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GTF2F2
- Gene Name
- general transcription factor IIF subunit 2
- Gene
- LOC101929259
- Gene Name
- uncharacterized LOC101929259
There are no transcripts associated with this gene.
View Full Product Details