Product Details

SNP ID
rs8032069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:100925936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTGAAAGGGTCAACAGGTTGGG[C/T]CTGAGAGGCCCACGTTTGTACCCTT
Phenotype
MIM: 600463 MIM: 610986
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALDH1A3 PubMed Links

Gene Details

Gene
ALDH1A3
Gene Name
aldehyde dehydrogenase 1 family member A3
There are no transcripts associated with this gene.

Gene
LOC101927751
Gene Name
uncharacterized LOC101927751
There are no transcripts associated with this gene.

Gene
LRRK1
Gene Name
leucine rich repeat kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024652.4 Intron NP_078928.3
XM_005254979.3 Intron XP_005255036.1
XM_011522012.2 Intron XP_011520314.1
XM_011522013.2 Intron XP_011520315.1
XM_011522014.2 Intron XP_011520316.1
XM_011522015.2 Intron XP_011520317.1
XM_011522016.2 Intron XP_011520318.1
XM_011522017.2 Intron XP_011520319.1
XM_011522018.2 Intron XP_011520320.1
XM_011522019.2 Intron XP_011520321.1
XM_017022570.1 Intron XP_016878059.1

View Full Product Details