Product Details

SNP ID
rs199748194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97357116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCAGGGTCGACGATCCTTTCTG[C/T]GGTTTTTGTGTCCCACCTGGGAACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RRP12 PubMed Links

Gene Details

Gene
RRP12
Gene Name
ribosomal RNA processing 12 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145114.1 4011 Missense Mutation CAC,CGC H1230R NP_001138586.1
NM_001284337.1 4011 Missense Mutation CAC,CGC H1191R NP_001271266.1
NM_015179.3 4011 Missense Mutation CAC,CGC H1291R NP_055994.2
XM_011539555.1 4011 Missense Mutation CAC,CGC H1263R XP_011537857.1
XM_011539556.2 4011 Intron XP_011537858.1

View Full Product Details