Product Details

SNP ID
rs199726601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:18659652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGCTCGTGATGGGGCTGATCTT[C/T]GCCAAACTGTGGAGCCTCTTCTGTA
Phenotype
MIM: 608909
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL5B PubMed Links

Gene Details

Gene
ARL5B
Gene Name
ADP ribosylation factor like GTPase 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178815.3 318 Silent Mutation TTC,TTT F5F NP_848930.1
XM_005252400.1 318 Silent Mutation TTC,TTT F5F XP_005252457.1
XM_005252401.4 318 Intron XP_005252458.1
Gene
NSUN6
Gene Name
NOP2/Sun RNA methyltransferase family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182543.3 318 Intron NP_872349.1
XM_011519383.1 318 Intron XP_011517685.1
XM_011519384.1 318 Intron XP_011517686.1
XM_011519385.1 318 Intron XP_011517687.1
XM_011519386.2 318 Intron XP_011517688.1
XM_011519387.2 318 Intron XP_011517689.1
XM_011519389.2 318 Intron XP_011517691.1
XM_017015906.1 318 Intron XP_016871395.1
XM_017015907.1 318 Intron XP_016871396.1
XM_017015908.1 318 Intron XP_016871397.1
XM_017015909.1 318 Intron XP_016871398.1
XM_017015910.1 318 Intron XP_016871399.1
XM_017015911.1 318 Intron XP_016871400.1
XM_017015912.1 318 Intron XP_016871401.1
XM_017015913.1 318 Intron XP_016871402.1
XM_017015914.1 318 Intron XP_016871403.1

View Full Product Details