Product Details

SNP ID
rs199602842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:7796143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGATTCAAGTTCGAAATATGGCA[A/G]CTTTGAAAGATAGTAAGTATGTTGT
Phenotype
MIM: 108729 MIM: 601720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP5C1 PubMed Links

Gene Details

Gene
ATP5C1
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001973.2 Intron NP_001001973.1
NM_001320886.1 Intron NP_001307815.1
NM_005174.3 Intron NP_005165.1
XM_017016290.1 Intron XP_016871779.1
Gene
KIN
Gene Name
Kin17 DNA and RNA binding protein
There are no transcripts associated with this gene.

View Full Product Details