Product Details

SNP ID
rs199744104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:32907086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGCAATTTAGAGACCAGCTTTAC[A/G]TCCGTAGTTTGGATTCTTGAAATTA
Phenotype
MIM: 135630
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGB1 PubMed Links

Gene Details

Gene
ITGB1
Gene Name
integrin subunit beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002211.3 2389 Intron NP_002202.2
NM_033668.2 2389 Missense Mutation CGT,TGT R797C NP_391988.1
NM_133376.2 2389 Intron NP_596867.1
Gene
SNORA86
Gene Name
small nucleolar RNA, H/ACA box 86
There are no transcripts associated with this gene.

View Full Product Details