Product Details
- SNP ID
-
rs201179782
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:47324864 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATGCCATCTCCATAACTGCCACA[G/A]AGGACTTCCCCTTCCAGAAGCACAT
- Phenotype
-
MIM: 605120
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GDF2
PubMed Links
Gene Details
- Gene
- GDF2
- Gene Name
- growth differentiation factor 2
There are no transcripts associated with this gene.
View Full Product Details