Product Details

SNP ID
rs201254715
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:22209959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTGTCTTGTGGGCTGCTGGTCT[G/T]TGGGTTGTTTCTTGGGCGGGACATT
Phenotype
MIM: 613249
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
EBLN1 PubMed Links

Gene Details

Gene
EBLN1
Gene Name
endogenous Bornavirus-like nucleoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199938.1 25 Missense Mutation AAG,CAG K9Q NP_001186867.1

View Full Product Details