Product Details
- SNP ID
-
rs200490757
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:97459234 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGAGGTACTTACCACAGGGGTGG[A/G]CAGGCGAGTGAGAGCATGCATGCAC
- Phenotype
-
MIM: 614777
MIM: 616750
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MMS19
PubMed Links
Gene Details
- Gene
- MMS19
- Gene Name
- MMS19 homolog, cytosolic iron-sulfur assembly component
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001289403.1 |
3579 |
Missense Mutation |
CCC,TCC |
P942S |
NP_001276332.1 |
NM_001289404.1 |
3579 |
Missense Mutation |
CCC,TCC |
P826S |
NP_001276333.1 |
NM_001289405.1 |
3579 |
Missense Mutation |
CCC,TCC |
P985S |
NP_001276334.1 |
NM_022362.4 |
3579 |
Missense Mutation |
CCC,TCC |
P985S |
NP_071757.4 |
XM_005270035.2 |
3579 |
Missense Mutation |
CCC,TCC |
P827S |
XP_005270092.1 |
XM_005270041.1 |
3579 |
Missense Mutation |
CCC,TCC |
P784S |
XP_005270098.1 |
XM_006717944.3 |
3579 |
Missense Mutation |
CCC,TCC |
P984S |
XP_006718007.2 |
XM_006717945.2 |
3579 |
Missense Mutation |
CCC,TCC |
P621S |
XP_006718008.1 |
XM_011540062.1 |
3579 |
Missense Mutation |
CCC,TCC |
P784S |
XP_011538364.1 |
XM_011540063.2 |
3579 |
Missense Mutation |
CCC,TCC |
P570S |
XP_011538365.1 |
XM_017016515.1 |
3579 |
Missense Mutation |
CCC,TCC |
P1024S |
XP_016872004.1 |
XM_017016516.1 |
3579 |
Missense Mutation |
CCC,TCC |
P1023S |
XP_016872005.1 |
XM_017016517.1 |
3579 |
Missense Mutation |
CCC,TCC |
P981S |
XP_016872006.1 |
XM_017016518.1 |
3579 |
Missense Mutation |
CCC,TCC |
P941S |
XP_016872007.1 |
XM_017016519.1 |
3579 |
Missense Mutation |
CCC,TCC |
P926S |
XP_016872008.1 |
XM_017016520.1 |
3579 |
Missense Mutation |
CCC,TCC |
P887S |
XP_016872009.1 |
XM_017016521.1 |
3579 |
Missense Mutation |
CCC,TCC |
P886S |
XP_016872010.1 |
XM_017016522.1 |
3579 |
Missense Mutation |
CCC,TCC |
P827S |
XP_016872011.1 |
XM_017016523.1 |
3579 |
Missense Mutation |
CCC,TCC |
P827S |
XP_016872012.1 |
XM_017016524.1 |
3579 |
Missense Mutation |
CCC,TCC |
P826S |
XP_016872013.1 |
XM_017016525.1 |
3579 |
Missense Mutation |
CCC,TCC |
P621S |
XP_016872014.1 |
XM_017016526.1 |
3579 |
Missense Mutation |
CCC,TCC |
P621S |
XP_016872015.1 |
XM_017016527.1 |
3579 |
Missense Mutation |
CCC,TCC |
P621S |
XP_016872016.1 |
XM_017016528.1 |
3579 |
Missense Mutation |
CCC,TCC |
P621S |
XP_016872017.1 |
XM_017016529.1 |
3579 |
Missense Mutation |
CCC,TCC |
P621S |
XP_016872018.1 |
XM_017016530.1 |
3579 |
Missense Mutation |
CCC,TCC |
P621S |
XP_016872019.1 |
XM_017016531.1 |
3579 |
Missense Mutation |
CCC,TCC |
P621S |
XP_016872020.1 |
XM_017016532.1 |
3579 |
Missense Mutation |
CCC,TCC |
P621S |
XP_016872021.1 |
XM_017016533.1 |
3579 |
Missense Mutation |
CCC,TCC |
P620S |
XP_016872022.1 |
XM_017016534.1 |
3579 |
Missense Mutation |
CCC,TCC |
P569S |
XP_016872023.1 |
XM_017016535.1 |
3579 |
Missense Mutation |
CCC,TCC |
P569S |
XP_016872024.1 |
- Gene
- ZDHHC16
- Gene Name
- zinc finger DHHC-type containing 16
There are no transcripts associated with this gene.
View Full Product Details