Product Details

SNP ID
rs200777085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97451798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTCTAGTACAGCGCTGGCGCTA[C/T]GGCAAGGTCTGCCTGCGCTCCCTGC
Phenotype
MIM: 606493 MIM: 614777 MIM: 616750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC1 PubMed Links

Gene Details

Gene
EXOSC1
Gene Name
exosome component 1
There are no transcripts associated with this gene.

Gene
MMS19
Gene Name
MMS19 homolog, cytosolic iron-sulfur assembly component
There are no transcripts associated with this gene.

Gene
ZDHHC16
Gene Name
zinc finger DHHC-type containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287803.1 339 Silent Mutation TAC,TAT Y41Y NP_001274732.1
NM_001287804.1 339 Silent Mutation TAC,TAT Y41Y NP_001274733.1
NM_032327.3 339 Silent Mutation TAC,TAT Y41Y NP_115703.2
NM_198043.2 339 Silent Mutation TAC,TAT Y41Y NP_932160.1
NM_198044.2 339 Silent Mutation TAC,TAT Y41Y NP_932161.1
NM_198045.2 339 Silent Mutation TAC,TAT Y41Y NP_932162.1
NM_198046.2 339 Silent Mutation TAC,TAT Y41Y NP_932163.1
XM_006718021.1 339 Silent Mutation TAC,TAT Y41Y XP_006718084.1
XM_006718022.1 339 Silent Mutation TAC,TAT Y41Y XP_006718085.1
XM_006718023.1 339 Silent Mutation TAC,TAT Y41Y XP_006718086.1
XM_006718024.1 339 Silent Mutation TAC,TAT Y41Y XP_006718087.1
XM_006718025.1 339 Silent Mutation TAC,TAT Y41Y XP_006718088.1
XM_006718026.1 339 Silent Mutation TAC,TAT Y41Y XP_006718089.1
XM_017016766.1 339 Silent Mutation TAC,TAT Y41Y XP_016872255.1
XM_017016767.1 339 Silent Mutation TAC,TAT Y41Y XP_016872256.1
XM_017016768.1 339 Silent Mutation TAC,TAT Y41Y XP_016872257.1
XM_017016769.1 339 Silent Mutation TAC,TAT Y41Y XP_016872258.1
XM_017016770.1 339 Silent Mutation TAC,TAT Y41Y XP_016872259.1
XM_017016771.1 339 Silent Mutation TAC,TAT Y41Y XP_016872260.1
XM_017016772.1 339 Silent Mutation TAC,TAT Y41Y XP_016872261.1
XM_017016773.1 339 UTR 5 XP_016872262.1
XM_017016774.1 339 UTR 5 XP_016872263.1

View Full Product Details