Product Details

SNP ID
rs200975836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73772878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCGGATTACCCGCTGTCGCTGC[A/G]GTGGCTGCCCGGGACCGCCTATCTG
Phenotype
MIM: 608842 MIM: 616932 MIM: 607185
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHCHD1 PubMed Links

Gene Details

Gene
CHCHD1
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 1
There are no transcripts associated with this gene.

Gene
FUT11
Gene Name
fucosyltransferase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284194.1 704 Missense Mutation CAG,CGG Q182R NP_001271123.1
NM_173540.2 704 Missense Mutation CAG,CGG Q182R NP_775811.2
XM_006717656.3 704 Missense Mutation CAG,CGG Q182R XP_006717719.1
Gene
SEC24C
Gene Name
SEC24 homolog C, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004922.3 704 Intron NP_004913.2
NM_198597.2 704 Intron NP_940999.1
XM_011540380.2 704 Intron XP_011538682.1
XM_011540381.2 704 Intron XP_011538683.1
XM_017016967.1 704 Intron XP_016872456.1
XM_017016968.1 704 Intron XP_016872457.1

View Full Product Details