Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001242487.1 | 2303 | Silent Mutation | TAC,TAT | Y667Y | NP_001229416.1 |
NM_001242488.1 | 2303 | Silent Mutation | TAC,TAT | Y667Y | NP_001229417.1 |
NM_015037.3 | 2303 | Silent Mutation | TAC,TAT | Y667Y | NP_055852.2 |
XM_005269648.2 | 2303 | Silent Mutation | TAC,TAT | Y667Y | XP_005269705.1 |
XM_005269653.3 | 2303 | Silent Mutation | TAC,TAT | Y667Y | XP_005269710.1 |
XM_005269655.3 | 2303 | Silent Mutation | TAC,TAT | Y667Y | XP_005269712.1 |
XM_006717725.3 | 2303 | Silent Mutation | TAC,TAT | Y667Y | XP_006717788.1 |
XM_011539544.1 | 2303 | Silent Mutation | TAC,TAT | Y667Y | XP_011537846.1 |
XM_017015978.1 | 2303 | Silent Mutation | TAC,TAT | Y667Y | XP_016871467.1 |