Product Details

SNP ID
rs201807534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:112153577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCACCACAAAACTCAGAATATA[C/T]TCTAGAAGTTTTTGTCGGTTGCATT
Phenotype
MIM: 602395
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPAM PubMed Links

Gene Details

Gene
GPAM
Gene Name
glycerol-3-phosphate acyltransferase, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244949.1 2670 Silent Mutation GAA,GAG E820E NP_001231878.1
NM_020918.5 2670 Silent Mutation GAA,GAG E820E NP_065969.3
XM_005269998.1 2670 Silent Mutation GAA,GAG E820E XP_005270055.1

View Full Product Details