Product Details

SNP ID
rs199512613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:28023816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAACACGTTTGGCAAATCCAGAAT[G/T]TACGTCTGCAGTTAACGAACTGTTT
Phenotype
MIM: 611271
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIF18A PubMed Links

Gene Details

Gene
KIF18A
Gene Name
kinesin family member 18A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031217.3 1429 Missense Mutation AAT,CAT N847H NP_112494.3
XM_017018379.1 1429 Missense Mutation AAT,CAT N847H XP_016873868.1
XM_017018380.1 1429 Missense Mutation AAT,CAT N403H XP_016873869.1

View Full Product Details