Product Details

SNP ID
rs199505340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4450017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTTATATCTTTATTCTTCAGGC[A/G]GTTCTACTGCTTGCCTCTCAGGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52K2 PubMed Links

Gene Details

Gene
OR52K2
Gene Name
olfactory receptor family 52 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005172.2 723 Silent Mutation GCA,GCG A226A NP_001005172.2

View Full Product Details