Product Details
- SNP ID
-
rs200792273
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:829393 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGGGGTGGCGTCAGCCCAGGGAG[A/C]GAACCCCTGCAGGACTCCCGAGGAG
- Phenotype
-
MIM: 602243
MIM: 614177
MIM: 609059
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CD151
PubMed Links
Gene Details
- Gene
- CD151
- Gene Name
- CD151 molecule (Raph blood group)
There are no transcripts associated with this gene.
- Gene
- CRACR2B
- Gene Name
- calcium release activated channel regulator 2B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286606.1 |
1581 |
Missense Mutation |
GAG,GCG |
E104A |
NP_001273535.1 |
NM_173584.4 |
1581 |
Missense Mutation |
GAG,GCG |
E104A |
NP_775855.3 |
XM_011520011.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_011518313.1 |
XM_011520019.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_011518321.1 |
XM_017017581.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_016873070.1 |
XM_017017582.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_016873071.1 |
XM_017017583.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_016873072.1 |
XM_017017584.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_016873073.1 |
XM_017017585.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_016873074.1 |
XM_017017586.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_016873075.1 |
XM_017017587.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_016873076.1 |
XM_017017588.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_016873077.1 |
XM_017017589.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_016873078.1 |
XM_017017590.1 |
1581 |
Missense Mutation |
GAG,GCG |
E104A |
XP_016873079.1 |
XM_017017591.1 |
1581 |
Missense Mutation |
GAG,GCG |
E104A |
XP_016873080.1 |
XM_017017592.1 |
1581 |
Missense Mutation |
GAG,GCG |
E104A |
XP_016873081.1 |
XM_017017593.1 |
1581 |
Missense Mutation |
GAG,GCG |
E104A |
XP_016873082.1 |
XM_017017594.1 |
1581 |
Missense Mutation |
GAG,GCG |
E104A |
XP_016873083.1 |
XM_017017595.1 |
1581 |
Missense Mutation |
GAG,GCG |
E104A |
XP_016873084.1 |
XM_017017596.1 |
1581 |
Missense Mutation |
GAG,GCG |
E104A |
XP_016873085.1 |
XM_017017597.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_016873086.1 |
XM_017017598.1 |
1581 |
Missense Mutation |
GAG,GCG |
E168A |
XP_016873087.1 |
XM_017017599.1 |
1581 |
Missense Mutation |
GAG,GCG |
E104A |
XP_016873088.1 |
- Gene
- PNPLA2
- Gene Name
- patatin like phospholipase domain containing 2
There are no transcripts associated with this gene.
View Full Product Details