Product Details

SNP ID
rs200956441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124543463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAACAGAAATCTATTAAAGAGAG[G/T]TTAAAAAGGAAGAAGTACATGGGAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR8B12 PubMed Links

Gene Details

Gene
OR8B12
Gene Name
olfactory receptor family 8 subfamily B member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005195.1 192 Silent Mutation NP_001005195.1

View Full Product Details