Product Details
- SNP ID
-
rs200130401
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:35619645 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGTCCGGGCGGGGCGCTGGTCTTT[C/T]TGGACAATGAGGCGGGCTTGGTGCA
- Phenotype
-
MIM: 612206
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FJX1
PubMed Links
Gene Details
- Gene
- FJX1
- Gene Name
- four jointed box 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014344.3 |
1459 |
Silent Mutation |
CTG,TTG |
L337L |
NP_055159.2 |
View Full Product Details