Product Details

SNP ID
rs200312424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59364829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACAGCCATGGCTTATGATCGTT[A/G]TGCTGCCATTTGTAACCCCCTGCTC
Phenotype
MIM: 615702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5AN1 PubMed Links

Gene Details

Gene
OR5AN1
Gene Name
olfactory receptor family 5 subfamily AN member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004729.1 371 Missense Mutation TAT,TGT Y124C NP_001004729.1

View Full Product Details