Product Details

SNP ID
rs200114560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124117545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGTGGCCTACTCACCCTCCAC[C/T]GGGAGCCAGGTAAGCCTAATTTGTG
Phenotype
MIM: 602929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VWA5A PubMed Links

Gene Details

Gene
VWA5A
Gene Name
von Willebrand factor A domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130142.1 243 Missense Mutation CGG,TGG R12W NP_001123614.1
NM_014622.4 243 Missense Mutation CGG,TGG R12W NP_055437.2
NM_198315.2 243 Missense Mutation CGG,TGG R12W NP_938057.1
XM_011542828.2 243 Missense Mutation CGG,TGG R28W XP_011541130.1

View Full Product Details